Learn More
Memmert™ Iq/Oq Document With Device-Specific Works Test Data (1 Temperature Value), For Un160Pa
Product Code. 16364347 Shop All Memmert Products
Product Code. 16364347 Supplier Memmert™ Supplier No. UN160paD00127
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
Includes temperature distribution survey for one free-selectable temperature value.
  • IQ/OQ document with works test data for chamber
  • 9 measuring points (size 30), 27 measuring points (sizes 55 - 1060) to DIN 12880:2007-05 (further temperature values on demand), PQ check list as support for validation by customer
Specifications
Product Type IQ/OQ Documentation