Learn More
Memmert™ Iq/Oq Document W/Device-Specific Works Test Data (Temperature And Vacuum Value), For Vo29
Product Code. 16314547 Shop All Memmert Products
Product Code. 16314547 Supplier Memmert™ Supplier No. VO29D00117
 Restricted Item
Estimated Shipment: 05-11-2026
Includes OQ check list with works test data for one free-selectable temperature value.
  • Temperature distribution survey for 5 measuring points to DIN 12880:2007-05 as support for validation by customer (VO, VOcool only) valid for one thermo shelf
Specifications
Product Type OQ Checklist