Learn More
Memmert™ IQ/OQ Document with Test Data for SN260
Shop All Memmert Products
Click to view available options
:
For Use With: SN260
Description
- Includes temperature distribution survey with 9 temperature measuring points (size 30) and 27 measuring points (sizes 55 - 1060) to DIN 12880:2007-05 (further temperature values on demand)
- PQ check list as support for validation by customer
Specifications
Specifications
| For Use With (Equipment) | SN260 Sterilizer |
| Product Type | IQ/OQ Documentation |
| Includes | Test Data |