Learn More
Memmert™ IQ/OQ Document with Test Data for SF160plus
Product Code. 17548155 Shop All Memmert Products
Change view
Click to view available options
For Use With: SF160plus
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17548155 For Use With: SF160plus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17548155 Supplier Memmert™ Supplier No. SF160plusD00127
IQ/OQ document with temperature distribution survey for one free-selectable temperature value.
  • Includes temperature distribution survey with 9 temperature measuring points (size 30) and 27 measuring points (sizes 55 - 1060) to DIN 12880:2007-05 (further temperature values on demand)
  • PQ check list as support for validation by customer
Specifications
For Use With (Equipment) SF160plus Sterilizer
Product Type IQ/OQ Documentation
Includes Test Data