Learn More
Memmert™ IQ/OQ Document with Test Data for IN260
Artikelnummer. 17537545 Alle ansehen Memmert Produkte
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
IN260 Incubator
missing translation for 'unitSize'
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. For Use With (Equipment) unitSize
17537545 IN260 Incubator Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17537545 missing translation for 'mfr' Memmert™ missing translation for 'supplierNo' IN260D00127
 Restricted Item
Estimated Shipment: 26-08-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
IQ/OQ document with temperature distribution survey for one free-selectable temperature value.
  • Includes temperature distribution survey with 9 temperature measuring points (size 30) and 27 measuring points (sizes 55 - 1060) to DIN 12880:2007-05 (further temperature values on demand)
  • PQ check list as support for validation by customer
Spezifikation
For Use With (Equipment) IN260 Incubator
Product Type IQ/OQ Documentation
Includes Test Data