Learn More
Memmert™ IQ/OQ Document with Test Data for IF110plus
Product Code. 17556655 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
IF110plus Incubator
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17556655 IF110plus Incubator Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17556655 Supplier Memmert™ Supplier No. IF110plusD00127
 Restricted Item
Estimated Shipment: 20-10-2026
IQ/OQ document with temperature distribution survey for one free-selectable temperature value.
  • Includes temperature distribution survey with 9 temperature measuring points (size 30) and 27 measuring points (sizes 55 - 1060) to DIN 12880:2007-05 (further temperature values on demand)
  • PQ check list as support for validation by customer
Specifikationer
For Use With (Equipment) IF110plus Incubator
Product Type IQ/OQ Documentation
Includes Test Data
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.