Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data - CO2 and Temperature
Product Code. 17179924 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
ICO105
ICO150
ICO240
ICO50
Unit Size:
Each
Product Code. 17179924
Brand: Memmert™ ICO105D38898
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For one free-selectable CO2 and temperature value
  • Includes temperature distribution survey at Memmert
  • For 27 measuring points to DIN 12880:2007-05
  • PQ check list as support for validation by customer.
Specifications
For Use With (Equipment) ICO105
Product Type IQ/OQ Document with Device-specific Works Test Data