Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data - CO2, Humidity and Temperature
Shop All Memmert ProductsDescription
- Includes temperature distribution survey at Memmert
- For 27 measuring points to DIN 12880:2007-05
- PQ check list as support for validation by customer.
Specifications
Specifications
| For Use With (Equipment) | ICO240 |
| Product Type | IQ/OQ Document with Device-specific Works Test Data |