Learn More
Memmert™ IQ Document with Test Data for UN750
Artikelnummer. 17500386 Alle ansehen Memmert Produkte
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
UN750 Oven
missing translation for 'unitSize':
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. For Use With (Equipment) missing translation for 'unitSize'
17500386 UN750 Oven Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17500386 missing translation for 'mfr' Memmert™ missing translation for 'supplierNo' UN750D00124
 Restricted Item
Estimated Shipment: 20-10-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
IQ document with device-specific works test data for validation by customer.
  • OQ/PQ check list as support for validation by customer
Spezifikation
For Use With (Equipment) UN750 Oven
Product Type IQ Document
Includes Test Data