missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Ipratropium-d3 Bromide (N-methyl-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | (±)-Ipratropium-d3 Bromide (N-methyl-d3) |
| Molecular Formula | C20 D3 H27 N O3 . Br |
| Quantity | 0.005 g |
| Synonym | 8-Isopropylatropinium-d3 bromide, Ipratropium-D3 (N-methyl-D3) bromide, Ipratropium-d3 bromide, (3-endo,8-syn)-3-(3-Hydroxy-1-oxo-2-phenylpropoxy)-8-(methyl-d3)-8-(1-methylethyl)-8-azoniabicyclo[3.2.1 ]octane bromide, 3α-Hydroxy-8-isopropyl-5αH-tropanium-d3 (±)-tropate bromide |
| InChI Formula | InChI=1 S/C20H30NO3.BrH/c1-14(2)21(3)16-9-10-17(21)12-18(11-16)24-20(23)19(13-22)15-7-5-4-6-8-15;/h4-8,14,16-19,22 H,9-13H2,1-3H3;1 H/q+1;/p-1/t16-,17+,18+,19?,21+;/i3D3; |
| SMILES | [Br-].[2 H]C([2 H])([2 H])[N@+]1(C(C)C)[C@@H]2 CC[C@H]1 C[C@H](C2)OC(=O)C(CO)c3ccccc3 |
| IUPAC Name | [(1 R,5 S)-8-propan-2-yl-8-(trideuteriomethyl)-8-azoniabicyclo[3.2.1 ]octan-3-yl] (2 RS)-3-hydroxy-2-phenylpropanoate;bromide |
| Molecular Weight (g/mol) | 415.3796 |
| Formula Weight | 414.1597 g/mol |
| Percent Purity | 99 atom % D, min 97% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction