Learn More
Ipazine, TRC
Specifications
Specifications
| Chemical Name or Material | Ipazine |
| CAS | 1912-25-0 |
| Molecular Formula | C10 H18 Cl N5 |
| Quantity | 1 g |
| Synonym | Ipazine, 1,3,5-Triazine-2,4-diamine, 6-chloro-N,N-diethyl-N'-(1-methylethyl)- (9CI), 6-Chloro-N2,N2-diethyl-N4-(1-methylethyl)-1,3,5-triazine-2,4-diamine (ACI), s-Triazine, 2-chloro-4-(diethylamino)-6-(isopropylamino)- (6CI, 8CI), 2-Chloro-4-(diethylamino)-6-(isop |
| InChI Formula | InChI=1S/C10H18ClN5/c1-5-16(6-2)10-14-8(11)13-9(15-10)12-7(3)4/h7H,5-6H2,1-4H3,(H,12,13,14,15) |
| SMILES | CCN(CC)c1nc(Cl)nc(NC(C)C)n1 |
| IUPAC Name | 6-chloro-2-N,2-N-diethyl-4-N-propan-2-yl-1,3,5-triazine-2,4-diamine |
| Molecular Weight (g/mol) | 243.736 |
| Formula Weight | 243.125 |