Learn More
Thermo Scientific Sure-Flow Ion-Selective Electrode, Fluoride
Product Code. 11780439 Shop All Cole-Parmer Products
Change view
Click to view available options
Connector Types:
BNC
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Unit Size
11780439 BNC Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11780439 Supplier Thermo Scientific Supplier No. 0572302
  • ONLINE EXCLUSIVE PRICE
    1660.00€
    1626.24€ / Pack of 1
    Save 33.76€
    2% Off
Estimated Shipment: 10-12-2026
The Fluoride Sure-Flow Ion-Selective Electrode is Orion-designed for optimum performance and the most comprehensive ISE selection.
  • Ionplus™ Sure-Flow™ Combination Ion-Selective Electrodes are easier to use than reference and half-cells—eliminating fumbling and tangles from multiple cables.
  • Combination Ion-Selective Electrodes feature both half-cell and reference cell in one electrode.
  • Half-Cell Ion-Selective Electrodes are the classic means of taking ion measurements.
  • Sure-flow™ technology provides stable, reproducible results.
  • The easy-to-clean design reduces the risk of inaccurate readings due to clogging of the reference junction.
  • All electrodes include a BNC connector, a 3 ft. cable, and manual.
  • Complete systems require-the purchase of a standardizing solution, filling solution for the reference electrode, ionic strength adjustor, and pH adjusters.
  • Chloride, cyanide, Sulfide, nitrate, and fluoride are approved for EPA methods for analysis of wastewater and for NPDES permitting.
Specifications
Connector Types BNC
Junction Sure-Flow Junction
Model 9609BNWP
Electrode Solid-State
Includes BNC connector, 3 ft. Cable, Manual
For Use With (Equipment) Thermo Scientific Orion Star™ Series pH Meters
Type Orion Sure-Flow ISE Probe