Learn More
Iodine Solution, 0.1M, Chem Lab™
Product Code. 11953163
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Product Code. 11953163
Brand: Chem-Lab CL05.1006.1000
Specifications
Quantity 1 L
Concentration or Composition (by Analyte or Components) 0.1mol/L
compliance-icons compliance-icons
Product Identifier
  • Iodine solution according HANUS 0.1 mol/l
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : iodo bromide