Learn More
Iodine Solution, 0.005M, Chem-Lab
Product Code. 10224582
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10224582 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10224582 Supplier Chem-Lab Supplier No. CL05.1001.1000
Used in laboratories for accurate volumetric analysis and titrations. Iodine Solution, 0.005M, Chem-Lab is a standardized solution with 1.269g I2 per 1L water. It is also used topically as a disinfectant and antibacterial agent. (Calibrate before use.)
Iodine is a nonmetallic element of the halogen group. It is a nutritionally essential element, especially important in thyroid hormone synthesis. In solution, it has anti-infective properties (when used topically).
Specifications
CAS 7553-56-2, 7732-18-5
Quantity 1 L
Molecular Formula I2
MDL Number MFCD00011355 MFCD00164163
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILES II
IUPAC Name diiodine
Molecular Weight (g/mol) 253.81
PubChem CID 807
ChEBI CHEBI:17606
Formula Weight 253.81g/mol
Concentration or Composition (by Analyte or Components) 0.005mol/L
Density 1.01g/mL