Learn More
Zeiss™ Invertoskop™ C Camera Accessories
Product Code. p-316732
Click to view available options
Type:
Video Adapter V25 C
Magnification Power:
1.0X
Diameter (English):
1 in.
For Use With (Equipment):
1 inch chip video camera
Unit Size:
Each
Product Code. 10706459
Brand: Zeiss™ 4512660000000
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For use with Carl Zeiss Invetoskop Microscopes
  • Video Adapter
  • V25 C
  • Available in 0.5 and 1.0 in. dia.
Specifications
Type Video Adapter V25 C
Color Black
Diameter (English) 1 in.
For Use With (Equipment) 1 inch chip video camera
Magnification Power 1.0X