Learn More
Zeiss™ Invertoskop™ C Camera Accessories
For use with Carl Zeiss Invetoskop Microscopes
Brand: Zeiss™ 000000-1108-963
Additional Details : Weight : 0.20000kg
Description
- Video Adapter
- V25 C
- Available in 0.5 and 1.0 in. dia.
Specifications
Video Adapter V25 C (3CCD) | |
1/2 in. | |
0.5X |
Black | |
0.5 inch chip video camera |