Learn More
Tork Intuition Sensor Skincare Dispenser S4
GREENER_CHOICE
Product Code. p-200113536 Shop All Tork Products
Click to view available options
Model:
Elevation
Image
Unit Size:
Each
Product Code. 30243791
Brand: Tork 460009
Only null left
Add to basket
Add to basket
Make a good impression on your guests with this dispenser that can be used with a wide range of Tork soaps, sanitisers and lotions in sealed bottles.
  • This easy-to-use design has sensor-driven, touch-free dispensing which reduces the risk of cross-contamination and promotes good hand hygiene for all users.
  • Help secure good hand hygiene - the factory-sealed bottle with a new pump for every refill helps reduce the risk of contamination and protect the formulations all the way to the hands.
  • Save staff time with effortless refilling.
  • High capacity reduces waste and helps reduce consumption by up to 50% (based on Essity test using Tork Foam Soap versus Tork Liquid Soap in Elevation dispenser).
TRUSTED_SUSTAINABILITY
Specifications
Product Type Sanitizer Dispenser
Color Silver
Alternate Part Number 7322540782318
Holds Hand Soap, Hand Sanitizer, Hand Lotion
Material Plastic, Metal
Mounting Wall Mount
No. of Compartments 1
Product Style Hinged Lid
Model Image