Learn More
STEMCELL Technologies IntestiCult™ Organoid Growth Medium (Mouse)
Product Code. 16656122
Encompass
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16656122 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16656122 Supplier STEMCELL Technologies Supplier No. 06005
 Restricted Item
Estimated Shipment: 08-10-2026
Cell culture medium for establishment and maintenance of mouse intestinal organoids
  • IntestiCult™ Organoid Growth Medium (Mouse) is a defined, serum-free cell culture medium for efficient establishment and long-term maintenance of mouse intestinal organoids
  • These organoids, or 'mini-guts', provide a convenient in vitro organotypic culture system for studying both the small and large intestinal epithelium and associated stem cell dynamics
  • Organoids grown in IntestiCult™ feature a polarized epithelium that contains all of the known cell types of the adult intestinal epithelium
  • Individual intestinal crypts rapidly form organoids when cultured in IntestiCult™ Organoid Growth Medium (Mouse)
  • Applications of these cultures include studying the development and function of the normal and tumorigenic intestinal epithelium, modeling intestinal disease, and investigating stem cell properties and regenerative therapy approaches
  • Organoid culture enables convenient in vitro characterization of a system with strong physiological relevance to the adult intestine
Specifications
Cell Type Intestinal Cells
For Use With (Application) Cell Culture; Differentiation; Expansion; Maintenance; Organoid Culture
Quantity 1 Kit