Learn More
STEMCELL Technologies IntestiCult™ Organoid Growth Medium (Human)
Product Code. 17189001
Encompass
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17189001 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17189001 Supplier STEMCELL Technologies Supplier No. 06010
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
Cell culture medium for establishment and maintenance of human intestinal organoids
  • Model key characteristics of the adult intestinal epithelium using intestinal organoids established and maintained with this complete medium formulation and optimized protocol
  • Using the easy-to-follow and robust protocol, you can derive organoids from human intestinal crypts in one week; organoid growth across donor samples, including those that are otherwise difficult to grow, is enabled by an enriched stem cell population
  • Organoids grown in IntestiCult Organoid Growth Medium (Human) incorporate a functional lumen enclosed by a polarized intestinal epithelial cell layer and, for versatile modeling applications, can be further differentiated in 3D or in 2D as submerged monolayers or air-liquid interface (ALI) cultures using IntestiCult Organoid Differentiation Medium (Human; Catalog #100-0214)
Specifications
Cell Type Intestinal Cells
For Use With (Application) Cell Culture; Differentiation; Expansion; Maintenance; Organoid Culture
Quantity 1 Kit