Learn More
Thermo Scientific Chemicals Intermedin, Human
Product Code. 15488239
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. 15488239
Brand: Thermo Scientific Chemicals J66760.M^
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Species Human
Purification Method HPLC
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 5102.80
Product Type Intermedin, human
Quantity 1 mg
Regulatory Status RUO - research use only