Learn More
BINDER™ Interior Lighting
Product Code. 16484449 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers ED 720
BINDER Chambers FD 720
BINDER Chambers FED 720
Unit Size:
Each
Product Code. 16484449
Brand: BINDER™ 80121824
Only null left
Add to basket
Add to basket
Interior Lighting - 400 V option model, with two 15 W light bulbs.
  • Includes two 15 W light bulbs
  • For use with assorted 400 V models of BINDER™ Drying and Heating Chambers
Specifications
Description Interior lighting - 400 V option model, with two 15 W light bulbs
For Use With (Equipment) BINDER Chambers ED 720
Type Interior Lighting
Includes 2 Bulbs
Voltage 400 V
Wattage 15 W