Learn More
Ebro Electronic™ SI 1100 Interface
Product Code. 11753549
Click to view available options
Interface:
 IF 100
Unit Size:
Each
Product Code. 11753549
Brand: Ebro Electronic™ 13406061
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
EBI IF 100 and Winlog.pro software.
  • With USB connector
  • Including antenna AL 111
  • With Winlog.pro™ software
  • Colored LED signals programming, readout and faulty execution
Order Info
Interface
TRUSTED_SUSTAINABILITY
Specifications
Type Datalogger System
Display Type No Display
Calibration Not Calibrated
Alarm Types None
Interface  IF 100
Model SI 1100
For Use With (Equipment) EBI 10, EBI 100 and EBI 11 data loggers