Learn More
WTW™ Interface Cable for PC
WTW offers printers with direct data output via a RS232 interface as well as suitable cables for the communication between devices and the PC
147.00€ - 158.00€
Specifications
Type | Interface Cable |
---|---|
For Use With (Application) | Used with Lab Station |
For Use With (Equipment) | For PC |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
10490822
|
WTW™
902842 |
6-pin Socket, 9/25 adapter |
158.00€
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
10002071
|
WTW™
902758 |
9-pin Socket |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
Interface Cable | |
Used with Lab Station | |
For PC |