Learn More
Eppendorf™ Interchangeable Platform for Innova™ 5000 Multi-tier Shaker
Product Code. 11399964
Change view
Click to view available options
Holds:
1 L Erlenmeyer Flasks
125 mL Erlenmeyer Flasks
2 L Erlenmeyer Flasks
2.8 L Fernbach Flasks
250 mL Erlenmeyer Flasks
4 L Erlenmeyer Flasks
500 mL Erlenmeyer Flasks
6 L Erlenmeyer Flasks
No. of Positions:
8
9
14
23
42
64
90
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds No. of Positions unitSize
11399964 500 mL Erlenmeyer Flasks 42 Each
11379964 125 mL Erlenmeyer Flasks 90 Each
11389964 250 mL Erlenmeyer Flasks 64 Each
11309974 1 L Erlenmeyer Flasks 23 Each
11319974 2 L Erlenmeyer Flasks 14 Each
11329974 2.8 L Fernbach Flasks 9 Each
11339974 4 L Erlenmeyer Flasks 9 Each
11349974 6 L Erlenmeyer Flasks 8 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11399964 Supplier Eppendorf™ Supplier No. M11969902
Add to basket
Add to basket
This interchangeable platform fits shaker model 5000.
  • Holds Erlenmeyer flasks or Fernbach flasks
  • Dedicated platform
TRUSTED_SUSTAINABILITY
Specifications
Holds 500 mL Erlenmeyer Flasks
For Use With (Equipment) Innova™ 5000 Multi-tier Shaker
No. of Positions 42