Description
- Aluminum construction provides fast heat-up
Specifications
Specifications
| For Use With (Equipment) | 10 Tubes |
| Material | Aluminum |
| No. of Places | 10 |
| Type | Block |
| For Use With (Application) | Falcon Tubes |
| Tube Size | 17 mm |
| Dimensions (L x W x H) | 140 x 50 x 63 mm |
| Depth (Metric) | 75 mm |