Learn More
Integra™ Miltex™ Standard Biopsy Punches
Product Code. 13490487
Encompass
Change view
Click to view available options
Diameter (Metric):
0.15 cm
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
13490487 0.15 cm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13490487 Supplier Integra™ Supplier No. 12460400
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Stainless-steel razor sharp cutting edge
  • Sealed in a sterile, peel-apart, see-through package
  • Sterilized by ethylene oxide
  • Single use
Order Info
Each box contains 50 sterile color-coded blister packs
Specifications
Blade Material Stainless Steel
Product Type Biopsy Punch
For Use With (Application) Offer the Convenience of Single-use Disposability without Sacrificing quality, Feel or Performance
Handle Type Ribbed
Description Seamless
Disposable Single-use
Sterility Sterile
Quantity 50/Pk.
Diameter (English) 0.05 in.
Diameter (Metric) 0.15 cm