Learn More
Thermo Scientific Chemicals Insulin B (22-25)
Product Code. p-5386225
Click to view available options
Quantity:
25mg
100mg
Unit Size:
100mg
25mg
Product Code. 15463789
Brand: Thermo Scientific Chemicals J61170.MB
Request Bulk or Custom Format
For research use
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Formulation Lyophilized powder
Quantity 25mg
Storage Requirements -30° to -10°C
Regulatory Status RUO
Protein Length H-Arg-gly-Phe-Phe-OH
Product Type Insulin B (22-25)
Recombinant Native
Form Lyophilized