Learn More
Julabo™ Insulation for Baths
Product Code. p-8002287
Click to view available options
Diameter (Metric):
12mm
14mm
23mm
Holds:
CR 12 mm ID, Reinf. 8 mm ID tubing
CR tubing 8 to 10 mm I.D.
Reinf.Tubing 12 mm/1/2 in. ID
Unit Size:
Each
Product Code. 10254011
Brand: Julabo™ 8930410
Only null left
Add to basket
Edge
Add to basket
Specifications
For Use With (Equipment) CF30, CF31, CF40, CF41 Cryo-compact circulator, ED-13 open heating bath circulator
Type Insulation
Holds CR tubing 8 to 10 mm I.D.
Size 1 m
Diameter (Metric) 14mm