Learn More
SHP Steriltechnik AG Instruments + Cooling Programme
Description
- Sterilisation of medical and laboratory instruments
- Controlled cooling to protect heat-sensitive materials
- Uniform steam penetration for reliable sterility
- Reproducible results for laboratory and clinical applications
Specifications
Specifications
| For Use With (Equipment) | Autoclaves |
| Product Type | Software |