Learn More
Hanna Instruments™ Shockproof Blue Rubber Boots
Product Code. 10024210
Change view
Click to view available options
Color:
Blue
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10024210 Blue Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10024210 Supplier Hanna Instruments™ Supplier No. HI710007
  • ONLINE EXCLUSIVE PRICE
    79.40€
    43.41€ / Pack of 1
    Save 35.99€
    45% Off
Estimated Shipment: 09-12-2026
A blue shockproof rubber boot designed to prevent damage in the case of accidental impact, is able to withstand harsh environmental conditions, and ensures users have a secure grip on their instrument when using it in the field, factory, or production room floor.
  • A blue shockproof rubber boot designed to protect your Hanna instrument and provide easier handling
  • Special boot helps prevent damage due to accidental impact
  • Durable rubber casing ensures users have a secure grip on their instrument when using it in the field, factory, or production room floor
  • Can withstand high temperatures of up to 200°C (392°F) as well as ozone and other environmental phenomena
  • Designed for select pH, multiparameter, and temperature meters
Specifications
Certifications/Compliance Rubber Boot, Shockproof, Blue, Ergo Style Casing, Designed to Protect your Instrument and Prevent Damage Due to Accidental Impact, Ozone and Environmental Phenomena
Color Blue
Product Type Rubber Boot