Learn More
Hanna Instruments™ Chlorine Reagent Sets, For Colorimetric Water Testing
Product Code. 11756003
Click to view available options
Range:
0 to 500 mg/L Total
0.00 to 10.00 mg/L Free and Total
0.00 to 5.00 mg/L Total
0.000 to 0.500 mg/L Free
No. of Tests:
100
300
600
For Use With (Application):
Chlorine Ultra High Range
Free Chlorine Ultra Low Range
Free and Total Chlorine High Range
Total Chlorine
Total Chlorine (DPD3)
Total Chlorine Ultra Low Range
Unit Size:
Each
Pack of 100
Product Code. 11756003
Brand: Hanna Instruments™ HI9577101
Only null left
Add to basket
Add to basket
Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Chlorine Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.
These high quality reagents are designed to follow accepted test methods for colorimetric determinations of total and/or free chlorine. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.
  • Pre-made, pre-measured water testing reagents for ease of use
  • Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
  • Clearly marked with expiration date and lot number on each packet and bottle for traceability
  • Reagent sets available for: Free and total chlorine (high range), Free chlorine (ultra low range), Total chlorine (ultra high range, ultra low range, undetermined range)
Specifications
For Use With (Application) Chlorine Ultra High Range
For Use With (Equipment) Compatible Hanna benchtop and portable photometers
Includes Individual packets
No. of Tests 100
Quantity 1 Set
Range 0 to 500 mg/L Total
Test Method Standard Methods, 20th edition, 4500-Cl
Type Water Testing Kit
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • X100 REACTIF CHLORE HR
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P334-IF ON SKIN: Immerse in cool water/wrap in wet bandages.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362-Take off contaminated clothing.
  • P404-Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contains : potassium iodide, potassium disulphate