Learn More
Abcam InstantBlue™ Coomassie Protein Stain (ISB1L)
Product Code. 32113177 Shop All Abcam Products
Change view
Click to view available options
Quantity:
30 mL
Unit Size:
30mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32113177 30 mL 30mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32113177 Supplier Abcam Supplier No. ab11921130mL
 Restricted Item
Estimated Shipment: 18-11-2026
InstantBlue™ (ISB1L) is a ready to use, ultra-fast and sensitive Coomassie Blue protein stain for polyacrylamide gels.
InstantBlue™ (ISB1L) is a ready to use, ultra-fast and sensitive Coomassie Blue protein stain for polyacrylamide gels.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 30 mL