Learn More
Abeba™ Insoles 3570
Product Code. 15620238
Click to view available options
Size:
40
41
42
43
44
45
46
Unit Size:
1 pair
Product Code. 15620238
Brand: Abeba™ 3570/43
Only null left
Add to basket
Add to basket
Massage knobs stimulate circulation. ESD dissipative and antistatic.
  • Microfiber
  • Anthracite
  • ESD dissipative according to DIN EN 61340
  • Antistatic
  • Stimulating circulation by massage knobs, comfortable orthopaedic footbed
  • Reflexor massage active-footbed
Specifications
Color Anthracite
Material Microfiber
Size 43
Style 3570