Learn More
Abeba™ Insoles 3569
Click to view available options
Size:
40
41
42
43
44
45
46
Unit Size:
1 pair
Description
- Microfiber
- ESD dissipative according to DIN EN 61340
- Antistatic
- Flat, smooth
- Footbed Reflexor
Specifications
Specifications
| Material | Microfiber |
| Size | 43 |
| Style | 3569 |