Learn More
Abeba™ Insoles 3551
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Description
- Gray
- Antistatic
- ESD dissipative according to DIN EN 61340
- Replaceable insole for shoes with air cushion sole
Specifications
Specifications
| Color | Gray |
| Size | 37 |
| Style | 3551 |