Learn More
Abeba™ Insoles 3551
Product Code. 15610128
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Product Code. 15610128
Brand: Abeba™ 3551/46
Only null left
Add to basket
Add to basket
Replaceable insole for shoes with air cushion sole is ESD dissipative and antistatic.
  • Gray
  • Antistatic
  • ESD dissipative according to DIN EN 61340
  • Replaceable insole for shoes with air cushion sole
Specifications
Color Gray
Size 46
Style 3551