Learn More
Buerkle™ V4A Stainless Steel Inserts for Novartos Multi
Product Code. 11817281 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
0 mL
0.5 mL
1 mL
1.5 mL
2 mL
missing translation for 'unitSize'
Each
Product Code. 11817281
missing translation for 'mfr': Buerkle™ 53580010
The V4A Stainless Steel Inserts for Buerkle™ Novartos Multi fit precisely and can easily be replaced. Inserts are available in volumes from 0 to 2.0 ml.
  • Fit precisely
  • Can easily be replaced
  • Stainless steel AISI 316L
TRUSTED_SUSTAINABILITY
Specifications
Product Type Insert
Material Stainless Steel (V4A)
Autoclavable Autoclavable
Capacity (Metric) 1 mL
Certifications/Compliance FDA
For Use With (Application) Novartos Multi Sampler