Learn More
Inosine-5'-monophosphate Disodium Salt Hydrate, TRC
Product Code. 30393111
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30393111 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30393111 Supplier Toronto Research Chemicals Supplier No. TRCI66500525G
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5'-Inosinic acid disodium salt hydrate
CAS 352195-40-5
Molecular Formula C10 H11 N4 O8 P . 2[Na] . x[H2 O]
Quantity 25 g
Synonym 5'-Inosinic acid, disodium salt, hydrate (9CI), 5'-Inosinic Acid Disodium Salt Hydrate
InChI Formula InChI=1S/C10H13N4O8P.2Na.H2O/c15-6-4(1-21-23(18,19)20)22-10(7(6)16)14-3-13-5-8(14)11-2-12-9(5)17;;;/h2-4,6-7,10,15-16H,1H2,(H,11,12,17)(H2,18,19,20);;;1H2/q;2*+1;/p-2/t4-,6-,7-,10-;;;/m1.../s1
SMILES O.[Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP(=O)([O-])[O-])n2cnc3C(=O)NC=Nc23
IUPAC Name disodium;[(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-1H-purin-9-yl)tetrahydrofuran-2-yl]methyl phosphate;hydrate
Molecular Weight (g/mol) 410.18
Formula Weight 410.0216