Learn More
Inosine-5'-monophosphate Disodium Salt Hydrate, TRC
Specifications
Specifications
| Chemical Name or Material | 5'-Inosinic acid disodium salt hydrate |
| CAS | 352195-40-5 |
| Molecular Formula | C10 H11 N4 O8 P . 2[Na] . x[H2 O] |
| Quantity | 50 g |
| Synonym | 5'-Inosinic acid, disodium salt, hydrate (9CI), 5'-Inosinic Acid Disodium Salt Hydrate |
| InChI Formula | InChI=1S/C10H13N4O8P.2Na.H2O/c15-6-4(1-21-23(18,19)20)22-10(7(6)16)14-3-13-5-8(14)11-2-12-9(5)17;;;/h2-4,6-7,10,15-16H,1H2,(H,11,12,17)(H2,18,19,20);;;1H2/q;2*+1;/p-2/t4-,6-,7-,10-;;;/m1.../s1 |
| SMILES | O.[Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP(=O)([O-])[O-])n2cnc3C(=O)NC=Nc23 |
| IUPAC Name | disodium;[(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-1H-purin-9-yl)tetrahydrofuran-2-yl]methyl phosphate;hydrate |
| Molecular Weight (g/mol) | 410.18 |
| Formula Weight | 410.0216 |