Learn More
Kartell™ Inoculation Loops and Needles
Manufactured with high impact polystyrene for good dimensional stability. Kartell™ Inoculation Loops and Needles provide a two–in–one sterile disposable tool for inoculation.
Brand:  Kartell™ 0265100
Product Code. 10040941
  • ONLINE EXCLUSIVE PRICE
    605.00€
    450.75€ / Pack of 8000
    Save 154.25€
    25% Off
Only null left
Estimated Shipment: 23-01-2026
Description
  • Gamma sterilized and packed in polybags
  • Looped end to retrieve an inoculum from a culture, transfer inoculum for streaking or transfer microscopic organisms
  • Needle end for picking off individual isolated colonies  or making stab inoculations into agar slants or tubes of solid culture medium
  • CE–IVD