Learn More
Heathrow Scientific™ Inoculating Needle/Loops
GREENER_CHOICE
Product Code. 16380776 Shop All Heathrow Scientific Products
Change view
Click to view available options
Volume:
1 μL
10 μL
Material:
ABS
Polypropylene
Unit Size:
Pack of 1000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume Material unitSize
16380776 1 μL Polypropylene Pack of 1000
16390776 1 μL ABS Pack of 1000
16300786 10 μL Polypropylene Pack of 1000
16310786 10 μL ABS Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16380776 Supplier Heathrow Scientific™ Supplier No. HS81121A
Only null left
Add to basket
Add to basket
Provide the choice to spread or pick colonies and cultures without the risk of contamination with Inoculating Needle/Loops.
  • Use loop end to uniformly streak a gel surface
  • Use needle end for removal of colonies
  • Lot-to-lot traceability
  • Sterile
  • Disposable
Specifications
Disposable Single-use
Volume 1 μL
Length (English) 7.9 in.
Length (Metric) 20 cm
Material Polypropylene
Sterility Sterile
Size Standard
Color Blue
Product Type Inoculating Loop and Needle, Flexible
Autoclavable Not Autoclavable
For Use With (Application) Spreading, Picking, and Transferring Colonies or Cultures