Learn More
Novus Biologicals™ 40nm Colloidal Gold Nanoparticles 10 OD
Product Code. 18765073 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
Unit Size:
100mL
10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18765073 100 mL 100mL
18706973 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
For Research Use Only
Product Code. 18765073 Supplier Novus Biologicals™ Supplier No. 2100100
  • ONLINE EXCLUSIVE PRICE
    4200.00€
    3971.10€ / 100mL
    Save 228.90€
    5% Off
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
For Research Use Only
  • Uniform Shape
  • Low co-efficient of variation in terms of size
  • High efficiency antibody binding for maximum sensitivity
  • Available at high concentrations to simplify assay development
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at 4°C. Do not freeze.
Quantity 100 mL