missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Can withstand the most extreme conditions (2.5M NaH at 70°C for ≈2 hours)
- Streptavidin is attached to InnovaCoat™ GOLD via an extended linker to facilitate high-affinity molecular interactions with avidin and streptavidin
Specifications
Specifications
| Content And Storage | Store at 4°C. Do not freeze. |
| Quantity | 1 mL |
| Product Type | Antibody Labeling Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?