Learn More
Novus Biologicals™ InnovaCoat (R) GOLD 40nm Mini Antibody Labeling Kit
Product Code. 18757863 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 Tests
3 Tests
Unit Size:
10 tests
3 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18757863 3 Tests 3 tests
18787093 10 Tests 10 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18757863 Supplier Novus Biologicals™ Supplier No. 2300005
Stock Up and Save This Summer
Use promo code 27950
View all promotions
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Antibody labeling kit
InnovaCoat nanoparticles in this kit are supplied as a freeze dried mixture. The conjugation reaction is initiated simply by adding a solution of the antibody, which becomes attached (via lysine residues) to the InnovaCoat surface. InnovaCoat GOLD can however be provided in a variety of formats (e.g. 10 OD liquid or as lyophilized nanoparticles) formulated to allow a one-step reaction with antibodies for bulk or custom orders. The kits are designed for research use and for the development of diagnostic products, specifically for the screening of potential antibodies or small scale proof-of-principle studies. The gold nanoparticles are also designed to be highly effective in lateral flow immunoassays.
Order Info
Quantity
TRUSTED_SUSTAINABILITY
Specifications
Product Type InnovaCoat GOLD 40nm Mini Antibody Labeling Kit
Content And Storage 3 Vials of InnovaCoat GOLD, 1 Vial of Reaction Buffer. Store at 4°C. Do not freeze.
Quantity 3 Tests