missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ InnovaCoat (R) GOLD - 40nm Midi Maleimide Antibody Labeling Kit
Shop All Bio Techne ProductsDescription
Features
- Proprietary surface coat
- Ultra-stable
- Covalently link antibodies and analytes
- Simple conjugation processes
- Different surface chemistries
- Enhanced assay sensitivity
- Protective coat prevents metal-antibody interactions
- Rapid screening of multiple antibodies for assay development
- Reduced antibody loading without aggregation
- No pH titrations
- Fully scalable – from R&D to production
- Custom InnovaCoat™ labeling services
Specifications
Specifications
| Content And Storage | -20°C |
| Quantity | 2.5 μg |
| Product Type | Antibody Labeling Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?