missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ InnovaCoat (R) Gold 40nm Biotin

Product Code. 18072084 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
200 μL
1 mL
Unit Size:
1mL
200µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18072084 200 μL 200µL
18025085 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only
Product Code. 18072084 Supplier Novus Biologicals™ Supplier No. 2400200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only

Gold nanoparticle with a unique protective surface coat. Covalently attached biotin ligand cannot desorb.

  • Can withstand the most extreme conditions (2.5M NaH at 70°C for ≈2 hours)
  • Biotin is attached to InnovaCoat™ GOLD via an extended linker to facilitate high-affinity molecular interactions with avidin and streptavidin
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C. Do not freeze.
Quantity 200 μL
Product Type Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.