Learn More
Eppendorf™ Innova™ 43R Incubator Shaker
Product Code. 11310025
Change view
Click to view available options
Orbit:
25 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Orbit Unit Size
11310025 25 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 11310025 Supplier Eppendorf™ Supplier No. M13200006
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Use these versatile incubator shakers for support high-speed applications and heavy workloads. New Brunswick Scientific™ Innova™ 43R Incubator Shakers handle large quantities or large volumes of cultures with precision and ease.
  • Console style shaker/incubator with triple-eccentric drive – provides large capacity shaking for flasks up to 6 liters
  • Programmable Innova controls – automatically changes temperature, speed, and optional photosynthetic and UV germicidal lights at timed intervals
  • Top mounted controls and display – for easy, ergonomic viewing and setting
  • Foot pedal release with easy opening lid – provides hands free operation
  • RS-232 port for data logging
  • Built-in water reservoir humidifies chamber to reduce sample evaporation, while also protecting unit from spills and including a drain for easy cleaning
TRUSTED_SUSTAINABILITY
Specifications
Orbit 25 mm
Speed Range 25 to 500 rpm
Electrical Requirements 230 V, 50 Hz
Voltage 230 V
Holds 6 L
Product Type Incubator Shaker
Frequency 50 Hz
Depth (Metric) 81.3 cm
Height (Metric) 165.1 cm
Width (Metric) 115.6 cm
Model Innova 43R
Timer Range 0.01 to 99:59 hr.