Learn More
3M™ Inner Protection Plates
Product Code. 11745294 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Can be used to increase one dark shade of welding filter
Can be used to increase two dark shades of welding filter
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11745294 Can be used to increase one dark shade of welding filter Pack of 5
11735304 Can be used to increase two dark shades of welding filter Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11745294 Supplier 3M™ Supplier No. 7100080227
  • ONLINE EXCLUSIVE PRICE
    58.00€
    43.99€ / Pack of 5
    Save 14.01€
    24% Off
 Restricted Item
Estimated Shipment: 20-01-2027
Specifications
Color Clear
Dimensions (L x W) 42 x 91 mm
For Use With (Equipment) Can be used to increase one dark shade of welding filter
For Use With (Application) Can be used to Increase one dark shade of welding filter.
Product Type Inner Protection Plate