Description
- Detachable
- Resuable
Specifications
Specifications
| Connector Types | S7-None |
| For Use With (Equipment) | Mettler Toledo InLab Sensors |
| For Use With (Application) | To connect Mettler Toledo InLab sensors to any meter |
| Length (English) Cable | 16.4 ft. |
| Length (Metric) Cable | 5.0 m |