Learn More
Mettler Toledo™ InLab™ Sensor Cables: S7 Connection
Description
- Detachable
- Resuable
Specifications
Specifications
| Connector Types | S7-BNC |
| For Use With (Equipment) | Mettler Toledo InLab Sensors |
| For Use With (Application) | To connect Mettler Toledo InLab sensors to any meter |
| Length (English) Cable | 9.8 ft. |
| Length (Metric) Cable | 3.0 m |